APO427944301 Apollo Scientific Ltd

2',4'-Dichloro-2-(4-fluorophenyl)acetophenone

  • MDL number MFCD16350604
  • Cas Number : 611220-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2,4-Dichlorophenyl)-2-(4-fluorophenyl)ethanone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63891

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63891
pictograms GHS05;GHS07
hstatements H315;H317;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart