APO427944332 Apollo Scientific Ltd

Methyl 3-bromo-6-chloro-2-fluorobenzoate

  • MDL number MFCD28040512
  • Cas Number : 1543224-54-9
  • Molecular Weight : 267.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-6-chloro-2-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-6-chloro-2-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart