APO427944338 Apollo Scientific Ltd

4'-Chloro-2-(3,4-difluorophenyl)acetophenone

  • MDL number MFCD23417249
  • Cas Number : 1373156-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63882

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63882
pictograms GHS05;GHS07
hstatements H317;H318
Availability details for
Quantity
  • Availability
  • Add to Cart