APO427944383 Apollo Scientific Ltd

4'-Chloro-2-(4-fluorophenyl)acetophenone

  • MDL number MFCD02063016
  • Cas Number : 126866-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Chlorophenyl)-2-(4-fluorophenyl)ethanone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63895

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63895
pictograms GHS05;GHS07
hstatements H317;H318
Availability details for
Quantity
  • Availability
  • Add to Cart