APO427944385 Apollo Scientific Ltd

2-(4-Chlorophenyl)-3'-fluoroacetophenone

  • MDL number MFCD12605188
  • Cas Number : 1183554-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4-Chlorophenyl)-1-(3-fluorophenyl)ethanone
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63902

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63902
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart