APO427944399 Apollo Scientific Ltd

2-(4-Chlorophenyl)-4'-methylacetophenone

  • MDL number MFCD11210879
  • Cas Number : 62006-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-114
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63900

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63900
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart