APO427944400 Apollo Scientific Ltd

2-(3-Fluorophenyl)-4'-methoxyacetophenone

  • MDL number MFCD11545169
  • Cas Number : 153529-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-75
Synonym 2-(3-Fluorophenyl)-1-(4-methoxyphenyl)ethanone
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63889

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63889
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart