APO427944413 Apollo Scientific Ltd

Methyl 2-acetamido-2-(dimethoxyphosphoryl)acetate

  • MDL number MFCD08443722
  • Cas Number : 89524-99-2
  • Molecular Weight : 239.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-acetamido-2-dimethoxyphosphorylacetate

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-acetamido-2-dimethoxyphosphorylacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart