APO427944432 Apollo Scientific Ltd

3'-Fluoro-2-(3-fluorophenyl)acetophenone

  • MDL number MFCD12605191
  • Cas Number : 40281-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49-51
Synonym 1,2-Bis(3-fluorophenyl)ethanone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63888

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63888
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart