APO427944434 Apollo Scientific Ltd

2-(3-Fluorophenyl)-4'-fluoroacetophenone

  • MDL number MFCD11210358
  • Cas Number : 370874-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(3-Fluorophenyl)-1-(4-fluorophenyl)ethanone
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63893

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63893
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart