APO427944435 Apollo Scientific Ltd

4'-Fluoro-2-(4-fluorophenyl)acetophenone

  • MDL number MFCD00055468
  • Cas Number : 366-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-98
Synonym 1,2-Bis(4-fluorophenyl)ethanone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 178-180/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63894

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63894
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart