APO427944447 Apollo Scientific Ltd

5-Bromo-2,4-dichloropyridine

  • MDL number MFCD09031035
  • Cas Number : 849937-96-8
  • Molecular Weight : 226.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-dichloropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-dichloropyridine
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart