APO427944448 Apollo Scientific Ltd

6-Bromo-tetral-1-one

  • MDL number MFCD04114378
  • Cas Number : 66361-67-9
  • Molecular Weight : 225.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 44-47
Iupac Name 6-bromo-3,4-dihydro-1(2H)-naphthalenone
Inchi Code 1S/C10H9BrO/c11-8-4-5-9-7(6-8)2-1-3-10(9)12/h4-6H,1-3H2
Inchi Key OSDHOOBPMBLALZ-UHFFFAOYSA-N

Description

Synonym 6-Bromo-3,4-dihydro-2H-naphthalen-1-one
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 44-47
Product Link https://store.apolloscientific.co.uk/product/6-bromo-tetral-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-tetral-1-one
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart