APO427944470 Apollo Scientific Ltd

Methyl 2,3,5-trifluoro-4-methoxybenzoate 1g pack

  • MDL number MFCD16877111
  • Cas Number : 1656263-44-3
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-235-trifluoro-4-methoxybenzoate

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart