APO427944481 Apollo Scientific Ltd

4-(Bromomethyl)-2-fluorobenzaldehyde

  • MDL number MFCD18378725
  • Cas Number : 1379307-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoro-4-formylbenzyl bromide
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63452

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63452
pictograms GHS05;GHS07
hstatements H302;H312;H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart