APO427944588 Apollo Scientific Ltd

2-Chloro-4-(difluoromethoxy)pyridine

  • MDL number MFCD13185848
  • Cas Number : 1206978-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63911

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63911
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart