APO427944590 Apollo Scientific Ltd

2-Chloro-6-(difluoromethoxy)pyridine

  • MDL number MFCD13185850
  • Cas Number : 1214377-45-3
  • Molecular Weight : 179.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-difluoromethoxypyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-difluoromethoxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart