APO427944646 Apollo Scientific Ltd

3-Chloro-N,N-diethylpropan-1-amine

  • MDL number MFCD00039385
  • Cas Number : 104-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-171/760mm
Density 0.89
Flash Point 59
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 82� 28 mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63668

Safety Information

signalword Danger
pcodes P210;P270
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63668
pictograms GHS02;GHS06
hstatements H226;H301;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart