APO428343616 Apollo Scientific Ltd

2-(Bromomethyl)-5-fluorobenzenesulphonyl chloride

  • MDL number MFCD28096593
  • Cas Number : 1820717-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Chlorosulphonyl)-4-fluorobenzyl bromide
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64197

Safety Information

signalword Danger
pcodes P232;P260;P280;P308+P313
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64197
pictograms GHS05
hstatements H314;EUH014
Availability details for
Quantity
  • Availability
  • Add to Cart