APO428343628 Apollo Scientific Ltd

4-(Bromomethyl)-2-fluorobenzenesulphonyl chloride

  • MDL number MFCD28096606
  • Cas Number : 1820684-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64213

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64213
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart