APO428343777 Apollo Scientific Ltd

2-(Bromomethyl)-3-fluorobenzonitrile

  • MDL number MFCD12025416
  • Cas Number : 635723-84-1
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-6-fluorobenzyl bromide, alpha-Bromo-3-fluoro-o-tolunitrile
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromomethyl-3-fluorobenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromomethyl-3-fluorobenzonitrile
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart