APO428343786 Apollo Scientific Ltd

3-(Bromomethyl)-5-fluorobenzonitrile

  • MDL number MFCD18394044
  • Cas Number : 853368-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-5-fluorobenzyl bromide
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64014

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64014
pictograms GHS05;GHS07
hstatements H302;H312;H332;H314;H317
Availability details for
Quantity
  • Availability
  • Add to Cart