APO428343813 Apollo Scientific Ltd

2-(Bromomethyl)-5-fluorobenzaldehyde

  • MDL number MFCD18205514
  • Cas Number : 945995-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-formylbenzyl bromide, alpha-Bromo-5-fluoro-o-tolualdehyde
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64043

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=64043
pictograms GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart