APO428343814 Apollo Scientific Ltd

Methyl 2-(bromomethyl)-5-chlorobenzoate

  • MDL number MFCD06804577
  • Cas Number : 668262-52-0
  • Molecular Weight : 263.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-2-(methoxycarbonyl)benzyl bromide, Methyl alpha-bromo-5-chloro-o-toluate
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 149-152/8mm
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromomethyl-5-chlorobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromomethyl-5-chlorobenzoate
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart