APO428343819 Apollo Scientific Ltd

Methyl 2-(bromomethyl)-3-iodobenzoate

  • MDL number MFCD27949192
  • Cas Number : 312747-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Iodo-6-(methoxycarbonyl)benzyl bromide
Purity 93
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64224

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64224
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart