APO428343825 Apollo Scientific Ltd

4-(Bromomethyl)-3-iodobenzoic acid

  • MDL number MFCD22491786
  • Cas Number : 496944-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Carboxy-2-iodobenzyl bromide, alpha-Bromo-3-iodo-p-toluic acid
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64229

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64229
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart