APO428343829 Apollo Scientific Ltd

Methyl 5-bromo-2-(bromomethyl)benzoate

  • MDL number MFCD11505949
  • Cas Number : 79670-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-(methoxycarbonyl)benzyl bromide, Methyl alpha,5-dibromo-o-toluate
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64231

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=64231
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart