APO428343843 Apollo Scientific Ltd

4-(Bromomethyl)-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD22492213
  • Cas Number : 859213-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-145
Synonym 2-(Bromomethyl)-5-carboxybenzotrifluoride, 4-Carboxy-2-(trifluoromethyl)benzyl bromide
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64207

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64207
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart