APO428497681 Apollo Scientific Ltd

1-Bromo-4-(isopropylthio)benzene

  • MDL number MFCD20484423
  • Cas Number : 70398-89-9
  • Molecular Weight : 231.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromo-4-isopropylthiobenzene

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/1-bromo-4-isopropylthiobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart