APO428497692 Apollo Scientific Ltd

5-Bromo-6-chloropicolinic acid

  • MDL number MFCD09910220
  • Cas Number : 959958-25-9
  • Molecular Weight : 236.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-6-chloropicolinic acid
Inchi Code 1S/C6H3BrClNO2/c7-3-1-2-4(6(10)11)9-5(3)8/h1-2H,(H,10,11)
Inchi Key SEBJRHGSFJEWFZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-6-chloropicolinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-6-chloropicolinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart