APO428497813 Apollo Scientific Ltd

Ethyl (4-iodophenyl)acetate

  • MDL number MFCD07779004
  • Cas Number : 15250-46-1
  • Molecular Weight : 290.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-4-iodophenylacetate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-4-iodophenylacetate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart