APO428497891 Apollo Scientific Ltd

3'-Bromo-5'-chloroacetophenone

  • MDL number MFCD13189467
  • Cas Number : 154257-85-9
  • Molecular Weight : 233.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-32
Synonym 1-(3-Bromo-5-chlorophenyl)ethanone
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-chloroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-chloroacetophenone
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart