APO428497936 Apollo Scientific Ltd

4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride

  • MDL number MFCD07774164
  • Cas Number : 874839-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=66207

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=66207
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart