APO428498055 Apollo Scientific Ltd

4'-Chloro-3'-fluoroacetophenone

  • MDL number MFCD04115858
  • Cas Number : 151945-84-5
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-42
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-fluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-fluoroacetophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart