APO428498210 Apollo Scientific Ltd

2-Methyl-3-(trifluoromethyl)benzylamine

  • MDL number MFCD06213551
  • Cas Number : 771572-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64978

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=64978
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart