APO428498223 Apollo Scientific Ltd

3,6-Dichloro-2-fluorobenzylamine

  • MDL number MFCD09025386
  • Cas Number : 916420-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-47
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64991

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64991
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart