APO428498304 Apollo Scientific Ltd

5-Chloro-2-(trifluoromethoxy)cinnamic acid

  • MDL number MFCD11519361
  • Cas Number : 1092461-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-138
Synonym 3-[5-Chloro-2-(trifluoromethoxy)phenyl]acrylic acid
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=65069

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=65069
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart