APO428498482 Apollo Scientific Ltd

3-Chloro-5-(trifluoromethyl)benzyl bromide

  • MDL number MFCD04115849
  • Cas Number : 886496-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=65249

Safety Information

signalword Danger
pcodes P310;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=65249
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart