APO428498544 Apollo Scientific Ltd

4-Fluoro-3,5-dimethylphenylacetic acid

  • MDL number MFCD09832375
  • Cas Number : 1000544-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=65311

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=65311
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart