APO428499039 Apollo Scientific Ltd

2-Fluoro-4-(trifluoromethoxy)benzonitrile

  • MDL number MFCD16652437
  • Cas Number : 1240256-78-3
  • Molecular Weight : 205.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethoxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethoxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart