APO428499400 Apollo Scientific Ltd

2-Chloro-5-fluoroisonicotinic acid

  • MDL number MFCD04972381
  • Cas Number : 884494-74-0
  • Molecular Weight : 175.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-fluoroisonicotinic acid
Inchi Code 1S/C6H3ClFNO2/c7-5-1-3(6(10)11)4(8)2-9-5/h1-2H,(H,10,11)
Inchi Key MEDPACAROROBNJ-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-fluoropyridine-4-carboxylic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluoroisonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluoroisonicotinic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart