APO428499503 Apollo Scientific Ltd

4-Bromo-2,3-difluorobenzylamine hydrochloride

  • MDL number MFCD12546123
  • Cas Number : 1980086-47-2
  • Molecular Weight : 258.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-2,3-difluorophenyl)methanamine hydrochloride
Inchi Code 1S/C7H6BrF2N.ClH/c8-5-2-1-4(3-11)6(9)7(5)10;/h1-2H,3,11H2;1H
Inchi Key SLPDOYCVDFSKKB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzylamine-hydrochloride

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzylamine-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart