APO428499528 Apollo Scientific Ltd

4-Bromo-2,3-difluorobenzyl alcohol

  • MDL number MFCD22493515
  • Cas Number : 162744-55-0
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Bromo-2,3-difluorophenyl)methanol
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-23-difluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart