APO429251033 Apollo Scientific Ltd

(4R)-2,2-Dioxido-4-isopropyl-1,2,3-oxathiazolidine, N-BOC protected

  • MDL number MFCD17018788
  • Cas Number : 1858273-22-9
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (4R)-4-isopropyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide
Inchi Code 1S/C10H19NO5S/c1-7(2)8-6-15-17(13,14)11(8)9(12)16-10(3,4)5/h7-8H,6H2,1-5H3/t8-/m0/s1
Inchi Key FWEXSIHWPAKSOP-QMMMGPOBSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-isopropyl-123-oxathiazolidine-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-isopropyl-123-oxathiazolidine-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart