APO429251037 Apollo Scientific Ltd

(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected

  • MDL number MFCD17018793
  • Cas Number : 1209467-60-6
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-phenyl-123-oxathiazolidine-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P304+P340;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/4r-22-dioxido-4-phenyl-123-oxathiazolidine-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart