APO429251073 Apollo Scientific Ltd

2-Cyano-3-phenoxy-5-(trifluoromethyl)pyridine

  • MDL number MFCD25970580
  • Cas Number : 1065607-70-6
  • Molecular Weight : 264.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-cyano-3-phenoxy-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-cyano-3-phenoxy-5-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart