APO429251144 Apollo Scientific Ltd

4-Fluoro-2-methoxyphenylacetonitrile

  • MDL number MFCD06660192
  • Cas Number : 886498-56-2
  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-fluoro-2-methoxyphenyl)acetonitrile
Inchi Code 1S/C9H8FNO/c1-12-9-6-8(10)3-2-7(9)4-5-11/h2-3,6H,4H2,1H3
Inchi Key RUIXXRPYOJHKQZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxyphenylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-methoxyphenylacetonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart