APO429251156 Apollo Scientific Ltd

3-Nitro-4-(trifluoromethoxy)benzoic acid

  • MDL number MFCD13185757
  • Cas Number : 784-77-0
  • Molecular Weight : 251.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitro-4-(trifluoromethoxy)benzoic acid
Inchi Code 1S/C8H4F3NO5/c9-8(10,11)17-6-2-1-4(7(13)14)3-5(6)12(15)16/h1-3H,(H,13,14)
Inchi Key GDBCQTKUSLDFRP-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-nitro-4-trifluoromethoxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-nitro-4-trifluoromethoxybenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart