APO429251378 Apollo Scientific Ltd

4-(Bromomethyl)-2-fluorobenzeneboronic acid

  • MDL number MFCD22573054
  • Cas Number : 1331945-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8°C
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromomethyl-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-bromomethyl-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart