APO429356968 Apollo Scientific Ltd

2-Bromo-3-fluoroiodobenzene

  • MDL number MFCD09056796
  • Cas Number : 851368-08-6
  • Molecular Weight : 300.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-fluoro-3-iodobenzene
Inchi Code 1S/C6H3BrFI/c7-6-4(8)2-1-3-5(6)9/h1-3H
Inchi Key MHMRKNHYHPORRR-UHFFFAOYSA-N

Description

Synonym 2-Bromo-1-fluoro-3-iodobenzene
Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroiodobenzene_PC51221

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-fluoroiodobenzene_PC51221
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart